Tisk…

4-(Difluoromethoxy)phenylacetic acid 98%

Dodavatel: Apollo Scientific
Varování

PC7690-1G PC7690-250MG PC7690-5G
APOSPC7690-1GEA 1910 CZK
APOSPC7690-1G APOSPC7690-250MG APOSPC7690-5G
4-(Difluoromethoxy)phenylacetic acid 98%
4-(Difluoromethoxy)phenylacetic acid
Vzorec: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00236239
CAS číslo: 243659-15-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR