Tisk…

2-Amino-4-(2,4-difluorophenylamino)-1,3,5-triazine

Dodavatel: Apollo Scientific
Varování

PC7655-1G
APOSPC7655-1GEA 2980 CZK
APOSPC7655-1G
2-Amino-4-(2,4-difluorophenylamino)-1,3,5-triazine
2-Amino-4-(2,4-difluorophenylamino)-1,3,5-triazine
Vzorec: C₉H₇F₂N₅
MW: 223,182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00104897
CAS číslo: 66088-46-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR