Tisk…

2,4,6-Tribromo-3,5-difluoropyridine

Dodavatel: Apollo Scientific
Nebezpečí

PC7529-1G
APOSPC7529-1GEA 8100 CZK
APOSPC7529-1G
2,4,6-Tribromo-3,5-difluoropyridine
2,4,6-Tribromo-3,5-difluoropyridine
Vzorec: C₅Br₃F₂N
Bod tání: 105…111 °C
Storage Temperature: Ambient
CAS číslo: 30841-93-1
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR