Tisk…

4'-Bromo-2'-(trifluoromethyl)acetanilide

Dodavatel: Apollo Scientific

PC7415-1G PC7415-5G PC7415-25G
APOSPC7415-1GEA 1870 CZK
APOSPC7415-1G APOSPC7415-5G APOSPC7415-25G
4'-Bromo-2'-(trifluoromethyl)acetanilide
4'-Bromo-2'-(trifluoromethyl)acetanilide
Vzorec: C₉H₇BrF₃NO
MDL Number: MFCD00672936
CAS číslo: 29124-62-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR