Tisk…

2',3',4'-Trifluoroacetophenone 98%

Dodavatel: Apollo Scientific
Varování

PC7202D-5G PC7202D-1G PC7202D-25G
APOSPC7202D-5GEA 705 CZK
APOSPC7202D-5G APOSPC7202D-1G APOSPC7202D-25G
2',3',4'-Trifluoroacetophenone 98%
2',3',4'-Trifluoroacetophenone
Vzorec: C₈H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD00061231
CAS číslo: 243448-15-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR