Tisk…

3-Cyano-1-propylboronic acid pinacol ester

Dodavatel: Apollo Scientific

PC7178-25G PC7178-5G PC7178-1G
APOSPC7178-25GEA 34060 CZK
APOSPC7178-25G APOSPC7178-5G APOSPC7178-1G
3-Cyano-1-propylboronic acid pinacol ester
3-Cyano-1-propylboronic acid pinacol ester
Vzorec: C₁₀H₁₈BNO₂
Bod varu: 85 °C/0,5mmHg
Storage Temperature: Freezer
MDL Number: MFCD09953480
CAS číslo: 238088-16-9
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR