Tisk…

4-(2-Fluorophenylcarbamoyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-((2-Fluorophenyl)carbamoyl)phenyl)boronic acid

PC6986-5G
APOSPC6986-5GEA 3060 CZK
APOSPC6986-5G
4-(2-Fluorophenylcarbamoyl)phenylboronic acid 98%
4-(2-Fluorophenylcarbamoyl)phenylboronic acid
Vzorec: C₁₃H₁₁BFNO₃
MW: 259.0408 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027205
CAS číslo: 874288-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR