Tisk…

1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether 98%

Dodavatel: Apollo Scientific
Varování

PC6787-5G
APOSPC6787-5GEA 3940 CZK
APOSPC6787-5G
1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether 98%
1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether
Vzorec: C₃Cl₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD01320777
CAS číslo: 32778-09-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR