Tisk…

Potassium bromo(difluoro)acetate 97+%

Dodavatel: Apollo Scientific
Varování

PC6410-25G PC6410-5G
APOSPC6410-25GEA 12240 CZK
APOSPC6410-25G APOSPC6410-5G
Potassium bromo(difluoro)acetate 97+%
Potassium bromo(difluoro)acetate
Vzorec: C₂BrF₂O₂·K
Storage Temperature: Ambient
MDL Number: MFCD07784243
CAS číslo: 87189-16-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR