Tisk…

2-Amino-4-(trifluoromethyl)benzonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Amino-4-cyanobenzotrifluoride

PC6362-5G
APOSPC6362-5GEA 1700 CZK
APOSPC6362-5G
2-Amino-4-(trifluoromethyl)benzonitrile 98%
2-Amino-4-(trifluoromethyl)benzonitrile
Vzorec: C₈H₅F₃N₂
MW: 186.1339 g/mol
Bod tání: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD04974124
CAS číslo: 1483-54-1
EINECS: 216-047-7
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR