Tisk…

2-Fluoro-3-(trifluoromethoxy)benzylamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: [2-Fluoro-3-(trifluoromethoxy)phenyl]methylamine

PC6357-1G PC6357-250MG PC6357-100MG
APOSPC6357-1GEA 10420 CZK
APOSPC6357-1G APOSPC6357-250MG APOSPC6357-100MG
2-Fluoro-3-(trifluoromethoxy)benzylamine
2-Fluoro-3-(trifluoromethoxy)benzylamine
Vzorec: C₈H₇F₄NO
MW: 209.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026479
CAS číslo: 1159512-65-8
UN: 2735
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR