Tisk…

Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC6220K-25G PC6220K-5G PC6220K-1G
APOSPC6220K-25GEA 4570 CZK
APOSPC6220K-25G APOSPC6220K-5G APOSPC6220K-1G
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride 97%
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride
Vzorec: C₁₂F₂₄O₄
Bod varu: 158…161 °C
Hustota: 1,8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00084743
CAS číslo: 27639-98-1
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR