Tisk…

3-Fluoro-5-(trifluoromethyl)cinnamic acid

Dodavatel: Apollo Scientific
Varování

PC5867-1G PC5867-5G
APOSPC5867-1GEA 622 CZK
APOSPC5867-1G APOSPC5867-5G
3-Fluoro-5-(trifluoromethyl)cinnamic acid
3-Fluoro-5-(trifluoromethyl)cinnamic acid
Vzorec: C₁₀H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00236296
CAS číslo: 575469-96-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR