Tisk…

Perfluoro(1,2,3-trichlorocyclopent-1-ene)

Dodavatel: Apollo Scientific

PC5793-1G PC5793-5G
APOSPC5793-1GEA 6050 CZK
APOSPC5793-1G APOSPC5793-5G
Perfluoro(1,2,3-trichlorocyclopent-1-ene)
Perfluoro(1,2,3-trichlorocyclopent-1-ene)
Vzorec: C₅Cl₃F₅
Storage Temperature: Ambient
MDL Number: MFCD00039452
CAS číslo: 3824-97-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR