Tisk…

3-Bromo-5-(difluoromethyl)benzoic acid 95%

Dodavatel: Apollo Scientific

PC57725-1G PC57725-250MG
APOSPC57725-1GEA 29760 CZK
APOSPC57725-1G APOSPC57725-250MG
3-Bromo-5-(difluoromethyl)benzoic acid 95%
3-Bromo-5-(difluoromethyl)benzoic acid
Vzorec: C₈H₅BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32070683
CAS číslo: 2169047-47-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR