Tisk…

Pentafluoropropionic anhydride 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA

PC5750-100G PC5750-25G
APOSPC5750-100GEA 10390 CZK
APOSPC5750-100G APOSPC5750-25G
Pentafluoropropionic anhydride 99%
Pentafluoropropionic anhydride

Useful derivatising agent of amines, alcohols, etc for Electron-Capture GC detection.

Vzorec: C₆F₁₀O₃
MW: 310.05
Bod varu: 73…74 °C
Hustota: 1.571 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000429
CAS číslo: 356-42-3
EINECS: 206-604-2
UN: 3265
ADR: 8 II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR