Tisk…

2-Bromo-4-chloro-5-fluorobenzyl bromide ≥95%

Dodavatel: Apollo Scientific

PC57123-250MG PC57123-1G PC57123-5G
APOSPC57123-250MGEA 5390 CZK
APOSPC57123-250MG APOSPC57123-1G APOSPC57123-5G
2-Bromo-4-chloro-5-fluorobenzyl bromide ≥95%
2-Bromo-4-chloro-5-fluorobenzyl bromide
Vzorec: C₇H₄Br₂ClF
MDL Number: MFCD30184439
CAS číslo: 1067882-61-4
UN: 3265

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR