Tisk…

4-Bromo-1-(difluoromethyl)-2-iodobenzene ≥95%

Dodavatel: Apollo Scientific

PC57120-250MG PC57120-1G
APOSPC57120-250MGEA 6850 CZK
APOSPC57120-250MG APOSPC57120-1G
4-Bromo-1-(difluoromethyl)-2-iodobenzene ≥95%
4-Bromo-1-(difluoromethyl)-2-iodobenzene
Vzorec: C₇H₄BrF₂I
MDL Number: MFCD18391114
CAS číslo: 1261671-69-5
UN: 3261

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR