Tisk…

Methyl 2-bromo-6-fluoronicotinate 95%

Dodavatel: Apollo Scientific

PC57105-1G PC57105-250MG
APOSPC57105-1GEA 24800 CZK
APOSPC57105-1G APOSPC57105-250MG
Methyl 2-bromo-6-fluoronicotinate 95%
Methyl 2-bromo-6-fluoronicotinate
Vzorec: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD14698113
CAS číslo: 1214385-74-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR