Tisk…

Methyl 2-bromo-5-fluoroisonicotinate 95%

Dodavatel: Apollo Scientific
Varování

PC57012-250MG PC57012-1G PC57012-5G
APOSPC57012-250MGEA 622 CZK
APOSPC57012-250MG APOSPC57012-1G APOSPC57012-5G
Methyl 2-bromo-5-fluoroisonicotinate 95%
Methyl 2-bromo-5-fluoroisonicotinate
Vzorec: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD09951945
CAS číslo: 885588-14-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR