Tisk…

2,3-Dibromo-1,4-difluorobenzene 95+%

Dodavatel: Apollo Scientific
Varování

PC56328-1G PC56328-25G PC56328-5G
APOSPC56328-1GEA 622 CZK
APOSPC56328-1G APOSPC56328-25G APOSPC56328-5G
2,3-Dibromo-1,4-difluorobenzene 95+%
2,3-Dibromo-1,4-difluorobenzene
Vzorec: C₆H₂Br₂F₂
MW: 271.885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922620
CAS číslo: 179737-33-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR