Tisk…

N-Methylperfluorobutanesulphonamide 97%

Dodavatel: Apollo Scientific
Varování

PC56060-1G PC56060-250MG PC56060-5G PC56060-100MG
APOSPC56060-1GEA 14680 CZK
APOSPC56060-1G APOSPC56060-250MG APOSPC56060-5G APOSPC56060-100MG
N-Methylperfluorobutanesulphonamide 97%
N-Methylperfluorobutanesulphonamide
Vzorec: C₅H₄F₉NO₂S
Bod tání: 36…37 °C
Storage Temperature: Ambient
MDL Number: MFCD19707291
CAS číslo: 68298-12-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR