Tisk…

Pentafluorobenzyl acrylate 97%

Dodavatel: Apollo Scientific
Varování

PC5537-1G PC5537-5G PC5537-250MG
APOSPC5537-1GEA 5630 CZK
APOSPC5537-1G APOSPC5537-5G APOSPC5537-250MG
Pentafluorobenzyl acrylate 97%
Pentafluorobenzyl acrylate
Vzorec: C₁₀H₅F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD00042328
CAS číslo: 153614-61-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR