Tisk…

Methyl 6-bromo-2-fluoro-3-methylbenzoate ≥97%

Dodavatel: Apollo Scientific

PC540039-250MG
APOSPC540039-250MGEA 622 CZK
APOSPC540039-250MG
Methyl 6-bromo-2-fluoro-3-methylbenzoate ≥97%
Methyl 6-bromo-2-fluoro-3-methylbenzoate
Vzorec: C₉H₈BrFO₂
MW: 247,061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24848718
CAS číslo: 1437780-03-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR