Tisk…

3-(Trifluoromethylthio)benzaldehyde 95+%

Dodavatel: Apollo Scientific
Varování

PC5389-10G PC5389-1G PC5389-5G
APOSPC5389-10GEA 14250 CZK
APOSPC5389-10G APOSPC5389-1G APOSPC5389-5G
3-(Trifluoromethylthio)benzaldehyde 95+%
3-(Trifluoromethylthio)benzaldehyde
Vzorec: C₈H₅F₃OS
Storage Temperature: Refrigerator
MDL Number: MFCD00236335
CAS číslo: 51748-27-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR