Tisk…

5-Fluoro-3-methoxypyridin-2-amine 95+%

Dodavatel: Apollo Scientific
Varování

PC535040-250MG
APOSPC535040-250MGEA 5460 CZK
APOSPC535040-250MG
5-Fluoro-3-methoxypyridin-2-amine 95+%
5-Fluoro-3-methoxypyridin-2-amine
Vzorec: C₆H₇FN₂O
Storage Temperature: Ambient
MDL Number: MFCD18256739
CAS číslo: 1097264-90-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR