Tisk…

4-Bromo-5-fluoro-2-iodobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC53386-250MG PC53386-1G PC53386-5G
APOSPC53386-250MGEA 4370 CZK
APOSPC53386-250MG APOSPC53386-1G APOSPC53386-5G
4-Bromo-5-fluoro-2-iodobenzoic acid 95%
4-Bromo-5-fluoro-2-iodobenzoic acid
Vzorec: C₇H₃BrFIO₂
MW: 344.9 g/mol
MDL Number: MFCD30297369
CAS číslo: 1870155-80-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR