Tisk…

1-(4-Fluoro-2-nitrophenyl)piperidin-4-ol

Dodavatel: Apollo Scientific

PC5338-1G
APOSPC5338-1GEA 6050 CZK
APOSPC5338-1G
1-(4-Fluoro-2-nitrophenyl)piperidin-4-ol
1-(4-Fluoro-2-nitrophenyl)piperidin-4-ol
Vzorec: C₁₁H₁₃FN₂O₃
Bod tání: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD08692453
CAS číslo: 942474-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR