Tisk…

2-Fluoro-3-nitrobenzonitrile

Dodavatel: Apollo Scientific
Varování

PC53131-1G PC53131-5G PC53131-25G
APOSPC53131-1GEA 910 CZK
APOSPC53131-1G APOSPC53131-5G APOSPC53131-25G
2-Fluoro-3-nitrobenzonitrile
2-Fluoro-3-nitrobenzonitrile
Vzorec: C₇H₃FN₂O₂
MW: 166.109 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11849938
CAS číslo: 1214328-20-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR