Tisk…

1-Bromo-4-difluoromethoxy-2-fluorobenzene

Dodavatel: Apollo Scientific
Varování

PC53076-1G PC53076-5G PC53076-10G
APOSPC53076-1GEA 7440 CZK
APOSPC53076-1G APOSPC53076-5G APOSPC53076-10G
1-Bromo-4-difluoromethoxy-2-fluorobenzene
1-Bromo-4-difluoromethoxy-2-fluorobenzene
Vzorec: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD18390897
CAS číslo: 1242258-38-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR