Tisk…

3-Bromo-2-chloro-6-fluorophenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC53028-1G PC53028-5G
APOSPC53028-1GEA 13660 CZK
APOSPC53028-1G APOSPC53028-5G
3-Bromo-2-chloro-6-fluorophenacyl bromide
3-Bromo-2-chloro-6-fluorophenacyl bromide
Vzorec: C₈H₄Br₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD28155021
CAS číslo: 1805575-98-7
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR