Tisk…

2'-(Trifluoromethyl)-biphenyl-4-methanamine hydrochloride 97%

Dodavatel: Apollo Scientific

PC530057-1G PC530057-500MG
APOSPC530057-1GEA 18620 CZK
APOSPC530057-1G APOSPC530057-500MG
2'-(Trifluoromethyl)-biphenyl-4-methanamine hydrochloride 97%
2'-(Trifluoromethyl)-biphenyl-4-methanamine hydrochloride
Vzorec: C₁₄H₁₂F₃N·ClH
Storage Temperature: Ambient
MDL Number: MFCD31536877
CAS číslo: 472964-20-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR