Tisk…

(S)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 97%

Dodavatel: Apollo Scientific

PC530011-1G PC530011-500MG PC530011-5G
APOSPC530011-1GEA 9920 CZK
APOSPC530011-1G APOSPC530011-500MG APOSPC530011-5G
(S)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 97%
(S)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid
Vzorec: C₁₄H₁₈FNO₄
Storage Temperature: Ambient
MDL Number: MFCD03427919
CAS číslo: 479064-88-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR