Tisk…

4-Fluoro-3-(trifluoromethoxy)benzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

PC53001-1G PC53001-5G PC53001-10G PC53001-25G
APOSPC53001-1GEA 7080 CZK
APOSPC53001-1G APOSPC53001-5G APOSPC53001-10G APOSPC53001-25G
4-Fluoro-3-(trifluoromethoxy)benzyl alcohol 98%
4-Fluoro-3-(trifluoromethoxy)benzyl alcohol
Vzorec: C₈H₆F₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06660255
CAS číslo: 86256-18-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR