Tisk…

N-Benzyl-3-(trifluoromethoxy)-DL-phenylglycine 95%

Dodavatel: Apollo Scientific

PC52472-1G PC52472-250MG PC52472-5G
APOSPC52472-1GEA 14880 CZK
APOSPC52472-1G APOSPC52472-250MG APOSPC52472-5G
N-Benzyl-3-(trifluoromethoxy)-DL-phenylglycine 95%
N-Benzyl-3-(trifluoromethoxy)-DL-phenylglycine
Vzorec: C₁₆H₁₄F₃NO₃
Storage Temperature: Ambient
CAS číslo: 2514942-07-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR