Tisk…

2-Amino-2-(4-fluorophenyl)propanoic acid 95%

Dodavatel: Apollo Scientific

PC520838-1G PC520838-250MG
APOSPC520838-1GEA 5590 CZK
APOSPC520838-1G APOSPC520838-250MG
2-Amino-2-(4-fluorophenyl)propanoic acid 95%
2-Amino-2-(4-fluorophenyl)propanoic acid
Vzorec: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD02662600
CAS číslo: 312-44-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR