Tisk…

2-Chloro-3-methylbenzotrifluoride 95%

Dodavatel: Apollo Scientific

PC520793-1G PC520793-5G
APOSPC520793-1GEA 5330 CZK
APOSPC520793-1G APOSPC520793-5G
2-Chloro-3-methylbenzotrifluoride 95%
2-Chloro-3-methylbenzotrifluoride
Vzorec: C₈H₆ClF₃
MW: 194.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226661
CAS číslo: 74483-48-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR