Tisk…

1-Trifluoromethoxymethyl-cyclopropanecarboxylic acid 97%

Dodavatel: Apollo Scientific

PC520780-1G PC520780-250MG PC520780-500MG
APOSPC520780-1GEA 44970 CZK
APOSPC520780-1G APOSPC520780-250MG APOSPC520780-500MG
1-Trifluoromethoxymethyl-cyclopropanecarboxylic acid 97%
1-Trifluoromethoxymethyl-cyclopropanecarboxylic acid
Vzorec: C₆H₇F₃O₃
MW: 184.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23382132
CAS číslo: 1408279-84-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR