Tisk…

2-Hydroxy-3-fluoro-4-(trifluoromethyl)pyridine 97%

Dodavatel: Apollo Scientific

PC520732-1G PC520732-250MG
APOSPC520732-1GEA 18880 CZK
APOSPC520732-1G APOSPC520732-250MG
2-Hydroxy-3-fluoro-4-(trifluoromethyl)pyridine 97%
2-Hydroxy-3-fluoro-4-(trifluoromethyl)pyridine
Vzorec: C₆H₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD16611126
CAS číslo: 1227594-89-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR