Tisk…

2-Difluoromethylbenzoic acid methyl ester 95%

Dodavatel: Apollo Scientific

PC520720-10G PC520720-1G PC520720-5G
APOSPC520720-10GEA 27780 CZK
APOSPC520720-10G APOSPC520720-1G APOSPC520720-5G
2-Difluoromethylbenzoic acid methyl ester 95%
2-Difluoromethylbenzoic acid methyl ester
Vzorec: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16140192
CAS číslo: 1018678-48-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR