Tisk…

2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 96%

Dodavatel: Apollo Scientific

PC520698-1G PC520698-5G
APOSPC520698-1GEA 4300 CZK
APOSPC520698-1G APOSPC520698-5G
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 96%
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride
Vzorec: C₅H₉ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD14525479
CAS číslo: 193140-71-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR