Tisk…

2-(1,1-Difluoroethyl)-1,3-difluorobenzene 95%

Dodavatel: Apollo Scientific

PC520669-5G
APOSPC520669-5GEA 31480 CZK
APOSPC520669-5G
2-(1,1-Difluoroethyl)-1,3-difluorobenzene 95%
2-(1,1-Difluoroethyl)-1,3-difluorobenzene
Vzorec: C₈H₆F₄
Storage Temperature: Ambient
MDL Number: MFCD11845808
CAS číslo: 1138445-06-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR