Tisk…

3-(Difluoromethyl)pyridine 97%

Dodavatel: Apollo Scientific

PC520604-1G PC520604-5G
APOSPC520604-1GEA 5990 CZK
APOSPC520604-1G APOSPC520604-5G
3-(Difluoromethyl)pyridine 97%
3-(Difluoromethyl)pyridine
Vzorec: C₆H₅F₂N
Storage Temperature: Ambient
MDL Number: MFCD11226584
CAS číslo: 76541-44-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR