Tisk…

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%

Dodavatel: Apollo Scientific

PC520588-1G PC520588-5G PC520588-25G
APOSPC520588-1GEA 1370 CZK
APOSPC520588-1G APOSPC520588-5G APOSPC520588-25G
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Vzorec: C₇H₃BrF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD10687236
CAS číslo: 749875-16-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR