Tisk…

3-Amino-4-fluorophenylboronic acid pinacol ester 96%

Dodavatel: Apollo Scientific
Varování

PC52054-1G PC52054-5G
APOSPC52054-1GEA 1620 CZK
APOSPC52054-1G APOSPC52054-5G
3-Amino-4-fluorophenylboronic acid pinacol ester 96%
3-Amino-4-fluorophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₇BFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11044430
CAS číslo: 1003575-43-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR