Tisk…

1-(Difluoromethyl)-4-ethoxy-2-fluoro-benzene

Dodavatel: Apollo Scientific

PC520339-1G
APOSPC520339-1GEA 34060 CZK
APOSPC520339-1G
1-(Difluoromethyl)-4-ethoxy-2-fluoro-benzene
1-(Difluoromethyl)-4-ethoxy-2-fluoro-benzene
Vzorec: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD29767788
CAS číslo: 2149591-21-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR