Tisk…

1-(Difluoromethyl)-4-(4-methoxyphenoxy)benzene

Dodavatel: Apollo Scientific

PC520288-1G
APOSPC520288-1GEA 35710 CZK
APOSPC520288-1G
1-(Difluoromethyl)-4-(4-methoxyphenoxy)benzene
1-(Difluoromethyl)-4-(4-methoxyphenoxy)benzene
Vzorec: C₁₄H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29767772
CAS číslo: 2149601-35-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR