Tisk…

5-benzyloxy-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine

Dodavatel: Apollo Scientific

PC520134-1G
APOSPC520134-1GEA 103810 CZK
APOSPC520134-1G
5-benzyloxy-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
5-benzyloxy-3-(difluoromethyl)-1H-pyrrolo[2,3-c]pyridine
Vzorec: C₁₅H₁₂F₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD29767617
CAS číslo: 2149597-58-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR