Tisk…

Ethyl 5-amino-4-bromo-1-(4-fluorophenyl)pyrazole-3-carboxylate

Dodavatel: Apollo Scientific

PC520005-1G
APOSPC520005-1GEA 25990 CZK
APOSPC520005-1G
Ethyl 5-amino-4-bromo-1-(4-fluorophenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(4-fluorophenyl)pyrazole-3-carboxylate
Vzorec: C₁₂H₁₁BrFN₃O₂
MW: 328,137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135137
CAS číslo: 1427022-75-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR