Tisk…

5-Bromo-3-fluorobenzene-1,2-diamine 97%

Dodavatel: Apollo Scientific
Varování

PC5177-1G PC5177-250MG
APOSPC5177-1GEA 705 CZK
APOSPC5177-1G APOSPC5177-250MG
5-Bromo-3-fluorobenzene-1,2-diamine 97%
5-Bromo-3-fluorobenzene-1,2-diamine
Vzorec: C₆H₆BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD03094270
CAS číslo: 517920-69-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR